Analytical balances, verified models with EC-type approval certificate (-0CE), Cubis®
供应商: Sartorius
Sartorius Cubis
SARBMSA224S-1CE-DUEA 78704.5 CNY
SARBMSU224S-0CE-DU SARBMSE125P-1CE-DU SARBMSE324S-0CE-DU SARBMSU125P-0CE-DU SARBMSE125P-0CE-DI SARBMSE125P-1CE-DI SARBMSE324P-0CE-DU SARBMSU125P-0CE-DI SARBMSE125P-0CE-DU SARBMSA1202S-0CED0 SARBMSA225S-1CE-DI SARBMSA224S-0CE-DA SARBMSA225P-0CE-DA SARBMSU324S-0CE-DU SARBMSA225S-1CE-DA SARBMSA224S-0CE-DI SARBMSE225S-0CE-DU SARBMSA225P-0CE-DI SARBMSU124S-1CE-DU SARBMSU225S-0CE-DI SARBMSA224S-0CE-DU SARBMSA524P-1CE-DU SARBMSE14202S000D0 SARBMSA10202S0CED0 SARBMSE124S-1CE-DU SARBMSA14202S000D0 SARBMSU225P-0CE-DI SARBMSA324S-1CE-DU SARBMSE524S-0CE-DI SARBMSE524S-0CE-DU SARBMSA324P-0CE-DA SARBMSU125P-1CE-DI SARBMSE225S-1CE-DI SARBMSA324S-0CE-DA SARBMSU225P-0CE-DU SARBMSU225S-0CE-DU SARBMSA324S-1CE-DA SARBMSU124S-0CE-DU SARBMSA324S-0CE-DI SARBMSE225S-1CE-DU SARBMSE124S-0CE-DU SARBMSA5203S-1CEDE SARBMSU324S-0CE-DI SARBMSA225P-1CE-DI SARBMSA224S-1CE-DI SARBMSA124S-0CE-DU SARBMSA125P-0CE-DI SARBMSA224S-1CE-DU SARBMSA225S-0CE-DA SARBMSA12201S0CED0 SARBMSU324P-0CE-DU SARBMSA125P-0CE-DA SARBMSA124S-0CE-DA SARBMSE225P-0CE-DI SARBMSA225P-1CE-DA SARBMSE225P-0CE-DU SARBMSU224S-0CE-DI SARBMSA524S-1CE-DU SARBMSA224S-1CE-DA SARBMSA1203S-0CEDE SARBMSE224S-0CE-DU
Analytical balances, verified models with EC-type approval certificate (-0CE), Cubis®
天平
The Cubis® range represents a modular concept from Sartorius to allow the customer to choose the components that suit the application area that is of interest. For the customer, the modular concept delivers quality weighing together with a high degree of flexibility without having to invest in expensive equipment that has features that may never be required. These models are verified versions for use in legal metrology in the European Economic area, (-0CE suffix). Non verified models are listed separately.
  • Choice of draught shield: Manual (DU); automatic (DA); automatic with built-in de-ioniser for effective removal of static charges (DI)
  • Choice of leveling: Assisted (0CE); automatic (1CE)
  • Second-generation monolithic weigh cells are even more rugged than their predecessors, which allows for top loading semi-microbalances (5 figure) with 0 to 220 g weighing capability
  • All models have removable control units and displays
  • RS232 and USB interfaces are included, and additionally on MSA and MSU models an Ethernet interface with implemented web services, plus SD card reader are also included
  • Easy to upgrade separate modules
  • IsoCAL; fully automatic calibration and adjustment
Three different display and control units are available to cover all facets of laboratory applications and enable a range of options for balance operation. The customer can then add the appropriate weighing platform to the choice of control unit.
In the analytical range (⁴/₅ figure balances), the choices are not only the control unit and balance capability, but the type of draught shielding that will benefit the application.
MSE control unit:
Black and white basic backlit LCD
Ideal for straightforward high accuracy weighing.
MSU control unit:
High resolution black and white, graphical and tiltable display
APC (Advanced Pharma Compliance) to meet the stringent requirements of the pharmaceutical industry
Suitable for wide ranging complex weighing tasks and includes SD card reader for transfer of not only data but task configurations and settings from balance to balance or to PC
Q-Pan; compensation of off centre loading effects/errors
Q-Level; intelligent warning function to detect if balance is perfectly level and provide adjustment prompts for the user to correct and re-level the balance
MSA control unit:
High resolution colour configuable TFT, graphical and tiltable, touch screen display
APC (Advanced Pharma Compliance) to meet the stringent requirements of the pharmaceutical industry
Suitable for wide ranging complex weighing tasks and includes SD card reader for transfer of, not only data, but task configurations and settings from balance to balance or to PC
Q-Pan; compensation of off-centre loading effects/errors
Q-Level; intelligent warning function to detect if balance is perfectly level and provide adjustment prompts for the user to correct and re-level the balance.
Here is an example of model MSA124S-000-DI where the modules are described as follows:
MSA = colour touch screen applications display module
124S-000 = 120 g four decimal place weighing module, not verified for use in legal metrology
DI = automatic draught shield with built-in static de-ioniser