HPLC columns, Avantor® ACE® Generix™ C18 and C8
供应商: Avantor®
Avantor® ACE® Generix™
ACETGEN-5C8-1546EA 2500 CNY
ACETGEN-5C8-1546 ACETGEN-5C18-GD ACETGEN-5C8-GD ACETGEN-5C18-2546 ACETGEN-5C18-1546 ACETGEN-3C8-GD ACETGEN-5C8-2546
HPLC columns, Avantor® ACE® Generix™ C18 and C8
色谱柱
Colums with high purity, highly reproducible silica and ultra narrow particle size distribution for excellent column efficiency.
  • Highly stable bonded phase
  • Ultra robust HPLC column packing technology
  • Excellent column and silica batch reproducibility
Every column is individually tested and supplied with a test chromatogram.
Certifications: ISO9001 quality standard