Iron, 1000 µg/ml (0,10% w/v), BAKER INSTRA-ANALYZED® reagent

供应商: Avantor®
BAKER INSTRA-ANALYZED®
BAKR5764-04EA 0 CNY
BAKR5764-04
Iron, 1000 µg/ml (0,10% w/v), BAKER INSTRA-ANALYZED® reagent
标准品 Cationic Standards Iron Standards
Plasma standard.

  • Prepared from the highest purity raw material 99,999%
  • Assay value accurate within ±0,3% of the assay value
  • Certificate of analysis lists the trace impurity level in µg/ml for over 65 elements
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR