Nickel, AA Standard, 1000 µg/mL (0,10% w/v), BAKER INSTRA-ANALYZED®, J.T. Baker®
供应商: Avantor®
BAKER INSTRA-ANALYZED®
BAKR6461-04EA 0 CNY
BAKR6461-04
Nickel, AA Standard, 1000 µg/mL (0,10% w/v), BAKER INSTRA-ANALYZED®, J.T. Baker®
标准品 Cationic Standards Nickel Standards
Prepared by dissolution of nickel metal of 99,995% spectral purity.Themetal content of the solution is confirmed by titration with EDTA,traceable to NIST SRM 915.
  • Standardised with NIST standard chemicals