Zinc, AAS standards, BAKER INSTRA-ANALYZED®, J.T. Baker®
BAKR6474-04EA 0 CNY
BAKR6474-04
Zinc, AAS standards, BAKER INSTRA-ANALYZED®, J.T. Baker®
Prepared by dissolution of zinc metal of 99,99% spectral purity. The metal content of the solution is confirmed by tritation with EDTA, traceable to NIST SRM 915.
  • Standardised with NIST standard chemicals
Caution: 10,000 µg/ml= (1,00% w/v)= 10000 mg/L