pH Indicator Papers, DUAL-TINT, J.T.Baker®
供应商: Avantor®
BAKR2868-01EA 447 CNY
BAKR2868-01 BAKR2867-01 BAKR2869-01
pH Indicator Papers, DUAL-TINT, J.T.Baker®
试纸 PH指示剂带
DUAL-TINT pH papers offer easy-to-read, precise pH indication.
  • Refillable dispenser
These pH papers are in a rolled 17-foot strip, 9,525 mm wide in a refillable dispenser complete with colour scale. Two different indicator zones on one strip separated by a white center division guarantees clear colour gradations and reliable pH values.
订购须知 The papers are also available as refills for the dispenser in packs of three rolls.