8260B acetate mix
供应商: Restek
REST30489EA 0 CNY
REST30489
8260B acetate mix
标准品 环境分析标准
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
This mix includes methyl acetate and n-amyl acetate. Contains seven acetates, 2000 µg/ml each in P and T methanol
Contents: n-amyl acetate, butyl acetate, ethyl acetate, isopropyl acetate, methyl acetate, propyl acetate and vinyl acetate.