PCR plates, Eppendorf twin.tec® forensic DNA grade
供应商: Eppendorf
Eppendorf twin.tec®
EPPE0030129.610EA 975 CNY
EPPE0030129.610 EPPE0030129.644 EPPE0030129.636 EPPE0030129.628 EPPE0030129.601
PCR plates, Eppendorf twin.tec® forensic DNA grade
微孔板 PCR板
PP wells with PC plate deck and skirt. The rigidity of the plate makes it ideal for robotic handling and the PP wells have an inert, non-wetting surface that prevents DNA, RNA and enzymes from binding. The ultra thin walls ensure optimum heat transfer into the sample. All plates are autoclavable and certified free from DNase, RNase and human genomic DNA.