Eppendorf Tubes® amber, with screw cap, 5 ml
供应商: Eppendorf
EPPE0030122.330EA 1023 CNY
EPPE0030122.330
Eppendorf Tubes® amber, with screw cap, 5 ml
管子 微管
Eppendorf® amber tubes are designed for the preparation and storage of light-sensitive samples and reagents.
  • Screw cap provides optimal grip and user friendly handling
  • Effective protection from energy-rich light in the lower wavelength range
  • Better visibility of the filling level inside the tube enables full control during liquid handling steps and thus reduces the risk of contamination