Eppendorf Tubes®, 5 ml, Protein LoBind, with screw cap
供应商: Eppendorf
EPPE0030122.356EA 1275 CNY
EPPE0030122.356
Eppendorf Tubes®, 5 ml, Protein LoBind, with screw cap
管子 微管
Eppendorf® Protein LoBind tubes are specifically designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.
  • Eppendorf LoBind material ensures optimised sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimise the risk of sample interference
  • Lot-certified PCR clean purity grade: Free of human DNA, DNase, RNase and PCR inhibitors
  • Precise lid sealing to minimise evaporation