VWR®, Carboys, Narrow Mouth, Round, with Spigot
供应商: VWR
VWR®
VWRU10755-130EA 6463.95 CNY
VWRU10755-130 VWRU10755-118
VWR®, Carboys, Narrow Mouth, Round, with Spigot
carboy水瓶
Used for storing and dispensing solutions and media. Ideal for sterile water storage.
  • Non-sterile
  • Excellent chemical resistance
  • Clear graduation marks
  • Integrated shoulder handle
Carboys are well suited for use in temperatures ranging from –100 to 100 °C. Carboys are leak-proof on standard test conditions. Spigots are tied to the handle, and require assembly by hand for usage. No tools are necessary for assembly.
Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth.
Cap size - 83 mm
Certifications: Resin meets US FDA 21 CFR, USP Class VI.
Caution: Always disassemble spigot and cap from carboy before autoclaving.