VWR® Talon®, Heavy-duty lab-frames
供应商: VWR
VWR® Talon®
VWRU60078-008EA 0 CNY
VWRU60078-008
VWR® Talon®, Heavy-duty lab-frames
支撑架
These freestanding units are specifically designed for heavy-duty work such as pilot plant setups.
  • Floor-mounted unit for heavy-duty use
  • Strong but lightweight steel channel frame
  • Counter-weighted cast iron feet provide sturdy support
  • Feet may be bolted to the floor for added stability
Outside frame is constructed of a strong but lightweight steel channel. Interior lattice made of lab frame rods that use a horizontal channel connector; one end accommodates rod, other end fits into channel and locks tightly with a bolt.
Loosen the bolt and the connector slides easily along channel track for simple lattice adjustment. Entire apparatus is supported on sturdy, counter-weighted cast iron feet, which may be bolted to the floor for added stability in top-heavy installations.
Frame dimensions: 1219 H×1829 W mm
Overall dimensions: 1905 H×1829 W mm
订购须知 Each lab frame includes five 1220 mm rods, three 1829 mm rods, fifteen S-connectors, sixteen channel connectors, twenty-four clamping nuts, two cast iron feet, one frame kit, and one wrench.