VWR®, Beaker Tongs
供应商: VWR
VWR®
VWRU82027-370EA 1038.33 CNY
VWRU82027-370
VWR®, Beaker Tongs
钳子
Tongs feature a three-prong design and carbon steel, cadmium-plated construction.
Designed for the safe and simple handling of beakers and similar vessels from 50 to 2000 ml capacities.