Traceable full scale thermometers update readings every second and display them on a 10 mm LCD readout.
Magnetic back allows for placement on a metal surface (incubators, freezers, water baths, and more)
Stainless steel probe with 3 m long cable and piercing tip allows constant exposure to extreme temperatures at the end of the range.
Units also incorporate a flip-open easel for use on the lab bench and a spring fastener for clipping to any edge.
Dial depth: 14 mm
Dial Ø: 54 mm
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Delivery information: Supplied with 1,5 V silver-oxide battery for up to 18 months contiuous use.