可重复使用培养管,无裙边
供应商: VWR Collection
VWR®
VWRU10545-936EA 813.91 CNY
VWRU10545-936 VWRU10545-946
可重复使用培养管,无裙边
管子 培养管
High quality test tubes designed and constructed for optimal performance.
  • Fire-polished top
  • Without rim
  • Durable white ceramic enamel marking spot
  • Suitable for temperatures up to 500 °C
  • Chemical resistant
Made from ASTM 438 Type 1 Class A borosilicate 33 expansion glass and as per ASTM specification E982 Type 4.