VWR®, Channel connector
供应商: VWR
VWR® Talon®
VWRU60079-012EA 0 CNY
VWRU60079-012
VWR®, Channel connector
搅拌棒
Special connector fastens lab frame rod to 41 mm steel channels.
  • Easy to adjust
  • Securely fastens lab frame rod to steen channels
  • Available individually or in bulk
Loosen the connector or slide up or down for easy adjustment of rod position.