Centrifuge, Clinical 50
VWRU82013-800EA 8721 CNY
VWRU82013-800
Centrifuge, Clinical 50
The Clinical 50 centrifuge features a window in the lid for viewing the rotor. This basic unit features variable speed which is suitable for a variety of applications in the clinical setting or research laboratory.
  • For use in physicians' offices and small laboratories
  • Clear lid with safety interlock
A safety interlock cuts power to the centrifuge when the lid is opened. Optional adapters accommodate smaller tubes, including common blood draw tubes. The rotor accepts 15 ml (16×130 mm) and 10 ml (16×100 mm) tubes directly.
The rugged ABS housing adds durability to the centrifuge and is easy to clean.