VWR® NSX Turbo, Safety spectacles
供应商: VWR
VWR®
VWRU89187-986EA 401.7 CNY
VWRU89187-986
VWR® NSX Turbo, Safety spectacles
眼镜
Protective eyewear with wraparound lens offers stylish and secure personal protection, with an array of comfort features to assist the wearer throughout extended use.
  • Rubberised temple tips and integrated nosepiece for extra comfort
  • Fits comfortably with half-mask respirators
  • Impact-resistant, scratch-resistant polycarbonate lenses
  • Provides 99,9% UV protection
Super lightweight eyewear for men and women combines a sleek appearance with rugged durability. Fits comfortably with half mask respirators.
Certifications: ANSI Z87.1