The VWR® series of flammable freezers are designed for refrigerated storage of flammable and volatile materials in general laboratory work areas.
- Temperature controller
- Digital setting accuracy
- Precise temperature maintenance
- Independent operation for security
- Monitoring and alarms
No electrical components are located in the interior of the unit, and the electrical components of the compressor are sealed in a vapour-proof enclosure for additional safety.
Units are equipped with high/low temperature alarm, sensor error alarm, power failure alarm, alarm mute with ring back, remote contacts, alarm validation, resettable 1-point min/max history. The units feature a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies.
Certifications: Units meet NFPA and OSHA guidelines 45 and 70, and are UL and C-UL listed.