The VWR® performance series of flammable material freezers are designed for refrigerated storage of flammable or volatile materials in general laboratory work areas.
- Data acquisition
- 24/7 data logging
- 24-hour digital temp chart
Units are equipped with USB port for data transfer, 4 - 20 mA output, min/max temp history, digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies. No electrical components are located in the interior of the unit, and the electrical components of the compressor are sealed in a vapour-proof enclosure for additional safety.
Certifications: Units meet NFPA and OSHA guidelines 45 and 70, and are UL and C-UL listed.