VWR® Performance, Chromatography refrigerators with natural refrigerants
供应商: VWR
VWR®
VWRU10819-608EA 149830.56 CNY
VWRU10819-608 VWRU10819-612 VWRU10819-610
VWR® Performance, Chromatography refrigerators with natural refrigerants
冰箱
These performance series chromatography refrigerator units are purpose built for scientific and medical applications requiring superior temperature management and state of the art product security and data logging.
  • Adjustable operating temperature range: 1 to 10 °C
  • Sample simulated digital temperature display
  • Password-protected control panel
  • Audible and visual high/low temperature alarms
Units feature a digital microprocessor temperature controller that allows the precise temperature management necessary for critical samples and supplies. Additional safety features include power outage alarm, alarm mute with ring back, adjustable volume and swivel casters to add height.
Remote alarm dry contacts and an access port are provided to simplify connections to existing facility monitoring systems.
Units are supplied with a three-years parts and labour warranty, with an additional five-years warranty on compressor parts.
It features a natural refrigerant that is environmentally friendly. It reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates and UL, ASHRAE compliance guidelines.
Certifications: UL and C-UL listed.