Thes refrigerators feature a digital microprocessor temperature controller that allows the precise temperature management necessary for critical samples and supplies.
- Digital setting accuracy
- Independent operation for security
Real time sample simulated temperatures are constantly displayed and supported by audible/visual high and low temperature alarms, along with power outage and sensor error alarms. Remote alarm dry contacts and an access port are provided to simplify connections to existing facility monitoring systems. Height does not include 127 mm for casters.
Environmentally friendly, natural hydrocarbon (HC) refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates and UL, ASHRAE compliance guidelines.
Certifications: U/C-UL listed.
Delivery information: Units are supplied with a two-years parts and labour warranty, with an additional five-years warranty on compressor parts.