VWR®Standard Series, Freezers, hazardous location, explosion proof
供应商: VWR
VWR®
VWRU10819-916EA 173052 CNY
VWRU10819-916 VWRU10819-914
VWR®Standard Series, Freezers, hazardous location, explosion proof
冷柜 防爆冰箱
These units are designed for use in environments where volatile or explosive conditions could potentially exist.
They are not intended for common laboratory/room environments that are not designed to be explosion-proof. All electrical components are housed outside of the unit.
Certifications: Meet NFPA and OSHA guidelines for Hazardous Locations, Class 1, Division II, Group C & D. UL/C-UL listed.
Caution: Warning: Units must be hard-wired in conduit to 120 VAC power sources. Power cords with plugs are not acceptable.