VWR®, Examination Light, Flexible-Arm
供应商: VWR
VWR® Flexible Arm™
VWRU23609-166EA 2053.69 CNY
VWRU23609-166
VWR®, Examination Light, Flexible-Arm
检查灯
These lights are ideal for inspection, quality control, electronics, and sample examination.
  • Place light exactly where you need it
  • Easily maneuverable
Fingertip control raises, lowers, and pivots the perfectly balanced lamp wherever desired. Weighted metal base and spring counterbalance permit smooth, fluid movement with excellent stability. Beam closely resembles daylight for correct colour detection.
Certifications: UL listed.
Delivery information: Supplied with white reflector and a 35W halogen bulb.