These highly absorbent wipes are constructed from a naturally reticulated, hydrophilic, polyurethane foam.
- High abrasion resistance and conformable characteristics of UltraSOLV® allow it to clean rough, coarse and bead blasted surfaces that will snag traditional polyester wipes
- Low levels of particle generation, ionics, organics, lot-to-lot consistency and fibre-free construction
- Unsurpassed contamination removal properties
- Sponge-like properties allow for excellent pick up and release of fluids
- Excellent static dissipative and tribocharging properties allow for safe cleaning of static sensitive assemblies, such as wafer transfer chambers in vacuum tools
Using Foamtec's patented cleaning process and foam technology, UltraSOLV® wipes are ideal for cleaning critical tools and assemblies.
Suitable for use in Class 10, ISO 4, M2.5 cleanroom environments.