VWR®, Laboratory Freezer with Natural Refrigerant, Solid Door Stainless Steel
供应商: VWR
VWR®
VWRU10820-418EA 290956.03 CNY
VWRU10820-418
VWR®, Laboratory Freezer with Natural Refrigerant, Solid Door Stainless Steel
冷柜 实验室通用冰箱
Stainless steel freezer with solid door, featuring microprocessor temperature controllers that allow the precise temperature management necessary for critical cold storage.
  • Stainless steel construction
  • Up-to-the-minute air and sample simulated temperature information
  • Swivel casters (adds 127 mm to height; included in dimensions)
The temperature display and alarm module offers a comprehensive array of features, providing up-to-the-minute air and sample simulated temperature information, as well as the added notification features of the door ajar and power failure alarms.
Remote alarm contacts enable end users to tie these units into existing alarm systems to notify appropriate personnel of alarm events as soon as possible.
Certifications: UL/C-UL listed. Environmentally friendly, natural hydrocarbon (HC) refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates and UL, ASHRAE compliance guidelines.
订购须知 Two-year parts and labor warranty, five years on compressor parts.