VWR®, Pocket liquid-in-glass thermometer
供应商: VWR
VWR®
VWRU89095-702EA 183.55 CNY
VWRU89095-702
VWR®, Pocket liquid-in-glass thermometer
温度计 Analogue Thermometers
Featuring a closed metal case to protect from breakage, these total immersion thermometers are compact, 160 mm, making them ideal for field studies, waste water treatment and any on-the-go applications.
  • Mercury-free
  • Contain blue organic liquid against lead-free, white back glass
  • Tested and inspected by our US quality team
Certifications: Traceable to NIST.
Delivery information: Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.