VWR®, Gilley induction coil set
供应商: VWR
VWR®
VWRU470005-394EA 864.6 CNY
VWRU470005-394
VWR®, Gilley induction coil set
教育材料 物理教育材料 Electricity/Magnetism Learning Activities
An excellent set designed for demonstrating electromagnetism and induced currents.
The set includes two electromagnetic coils mounted on plastic saddle blocks, round and square iron cores, split round iron cores, a double core, and a card for showing lines of force.