Disposable particle filtering respirators, FFP3, uvex silv-Air 7315
供应商: Uvex
UVEX8707.311EA 6550 CNY
UVEX8707.311 UVEX8707.315
Disposable particle filtering respirators, FFP3, uvex silv-Air 7315
FFP3 masks are particularly suitable in welding and soldering, or when working with high concentrations of hazardous, microscopic respirable dust or with chemical compounds. All FFP3 respirators afford reliable protection against dust of up to 30-times the limit value (TLV) and all feature a valve.
  • Soft material edges
  • All-around sealing lip for optimal wearer comfort and secure fit
  • Four-point headband attachment for a comfortable fit
  • Without nose clip (metal free)
  • Low breathing resistance
Exhalation valve to further reduce breathing resistance, by helping to minimise moisture build up inside the mask. Ideal for construction and related industries.
*One size fits all
Certifications: EN 149:2001+A1:2009