VWR®, Magnetic stirrer, air operated turbine
供应商: VWR
VWR®
VWRU89049-012EA 1592.75 CNY
VWRU89049-012
VWR®, Magnetic stirrer, air operated turbine
搅拌器 磁力搅拌器
This compact, small-footprint stirrer only uses 206 - 551 mbar air pressure and is ideal for stirring vessels up to one litre.
  • Tubing is supplied to connect to an air supply
  • All parts are made from high-impact polystyrene
  • Ensured long life and easy cleanup in case of spills
The air-operated turbine magnetic stirrer can be used with magnetic stirring bars up to 250 mm in length.