Order Entry
Size exclusion chromatography columns, InfinityLab AdvanceBio
Size exclusion chromatography columns, InfinityLab AdvanceBio
Certificates
undefined
Size exclusion chromatography columns, InfinityLab AdvanceBio
Specifications
  • Catalog No:
  • HEWLPL1180-5301
  • HEWLPL1180-1301
  • Description:
  • 300 Å
  • 300 Å
  • Max. pressure:
  • 400 bar
  • 400 bar
  • pH range:
  • 2 – 8,5
  • 2 – 8,5
Specifications
Related Information
Product Information
  • Catalog #
    Supplier #
    Column Format
    Phase
    Column hardware
    Pore size
    Particle Size
    Ø int.
    Availability
    Price
  • HEWLPL1180-5301
    PL1180-5301
    Analytical
    SEC
    Stainless steel
    300 Å
    2,7 µm
    7,80 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件 - ¥0.00
    ¥0.00
    1 * 1 件
       ¥0.00
    Specifications:
    More
    Accessories
    Specifications
    Description
    300 Å
    Environmentally Preferable
    Max. pressure
    400 bar
    pH range
    2 – 8,5
  • HEWLPL1180-1301
    PL1180-1301
    Guard column
    SEC
    Stainless steel
    300 Å
    2,7 µm
    7,80 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件 - ¥0.00
    ¥0.00
    1 * 1 件
       ¥0.00
    Specifications:
    More
    Accessories
    Specifications
    Description
    300 Å
    Environmentally Preferable
    Max. pressure
    400 bar
    pH range
    2 – 8,5