PCR tubes,
VWRI732-3611EA 660.15 CNY
VWRI732-3611
PCR tubes,
Individual PCR tubes feature evaporation-resistant, easy-to-open-and-close lids that prevent sample loss.
  • Ultra-thin, uniform wells ensure optimal heat transfer and reaction efficiency
  • Available in ultra-clear, assorted, or specific colours
Certifications: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free.
Packaging: Conveniently packaged in reclosable bags to minimise risk of contamination.