DEC-GLASS®, Glass and Plexiglas® Cleaner
供应商: Veltek
DEC-GLASS®
VELTDG-03-16Z-EEA 4991.49 CNY
VELTDG-03-16Z-E
DEC-GLASS®, Glass and Plexiglas® Cleaner
清洗剂 洗涤剂和表面清洁剂
VAI manufactures a glass and plexiglass cleaner that is processed to comply with the standards required by pharmaceutical, biotechnology, medical device, and healthcare industries. DEC-GLASS® was developed for use in cleaning rotation cycles to remove residue from glass and plexiglass surfaces.
  • Filtered at 0.2 microns
  • Filled in ISO 5 (Grade A/B, Former Class 100)
  • Ready-to-use residue remover for glass and plexiglass
  • Sterile and formulated with water for injection
  • Designed for use pharmaceutical and biotechnology operations
An excellent cleaner designed for all washable, porous, and non-porous environmental surfaces. Using DEC-GLASS®, one can assure that noticeable and unnoticeable residues, smudges, oils, and dirt buildup are removed, thus returning the surface to its original form.
DEC-GLASS® is available in a sterile 16 oz trigger spray bottle that is terminally sterilized through gamma irradiation. It is delivered with lot specific documentation.