VAI manufactures a glass and plexiglass cleaner that is processed to comply with the standards required by pharmaceutical, biotechnology, medical device, and healthcare industries. DEC-GLASS® was developed for use in cleaning rotation cycles to remove residue from glass and plexiglass surfaces.
- Filtered at 0.2 microns
- Filled in ISO 5 (Grade A/B, Former Class 100)
- Ready-to-use residue remover for glass and plexiglass
- Sterile and formulated with water for injection
- Designed for use pharmaceutical and biotechnology operations
An excellent cleaner designed for all washable, porous, and non-porous environmental surfaces. Using DEC-GLASS®, one can assure that noticeable and unnoticeable residues, smudges, oils, and dirt buildup are removed, thus returning the surface to its original form.
DEC-GLASS® is available in a sterile 16 oz trigger spray bottle that is terminally sterilized through gamma irradiation. It is delivered with lot specific documentation.