DL-Alanylglycine

供应商: TCI
TCIAA0183-1GEA 810 CNY
TCIAA0183-1G
DL-Alanylglycine
蛋白质
  • Melting point: 225 °C
  • Solubility in water: Almost transparency

Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-DL-Ala-Gly-OH
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR