1-Allyl-3-(2-hydroxyethyl)-2-thiourea ≥98.0% (以总氮计)
供应商: TCI EUROPE
|
Warning
|
TCIAA0230-25GEA
0
CNY
TCIAA0230-25G
TCIAA0230-500G
1-Allyl-3-(2-hydroxyethyl)-2-thiourea ≥98.0% (以总氮计)
1-Allyl-3-(2-hydroxyethyl)-2-thiourea
分子式:
C₆H₁₂N₂OS 分子量: 160,24 g/mol |
MDL编号:
MFCD00002838 CAS号: 105-81-7 EINECS: 203-334-7 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Solubility in hot Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...