1-Amino-8-naphthol-4-sulphonic acid ≥90.0% (以总氮计)
供应商: TCI EUROPE
|
Warning
|
TCIAA0369-25GEA
0
CNY
TCIAA0369-25G
1-Amino-8-naphthol-4-sulphonic acid ≥90.0% (以总氮计)
1-Amino-8-naphthol-4-sulphonic acid
分子式:
C₁₀H₉NO₄S 分子量: 239,25 g/mol |
MDL编号:
MFCD00003992 CAS号: 83-64-7 EINECS: 201-491-6 |
产品技术规格
Appearance | Light gray to Brown to Black powder to crystal |
Purity (with Total Nitrogen) | min. 90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...