3-Nitroaniline-4-sulphonic acid ≥94.0% (通过HPLC)

供应商: TCI EUROPE

TCIAA0373-25GEA 945 CNY
TCIAA0373-25G
3-Nitroaniline-4-sulphonic acid ≥94.0% (通过HPLC)
3-Nitroaniline-4-sulphonic acid
分子式: C₆H₆N₂O₅S
分子量: 218,18 g/mol
MDL编号: MFCD00059185
CAS号: 712-24-3
EINECS: 211-920-9

现在订购

产品技术规格

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 94.0 area%
Purity (Neutralization titration) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR