2-Aminophenol hydrochloride ≥98.0% (通过HPLC,总氮)

Warning

TCIAA0391-5G
2-Aminophenol hydrochloride ≥98.0% (通过HPLC,总氮)
2-Aminophenol hydrochloride
分子式: C₆H₇NO
分子量: 145,59 g/mol
存储条件: Refrigerator
MDL编号: MFCD00035471
CAS号: 51-19-4

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR