Anthraquinone-1,5-disulphonic acid disodium salt ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAA0506-25GEA 0 CNY
TCIAA0506-25G
Anthraquinone-1,5-disulphonic acid disodium salt ≥98.0% (通过滴定分析)
Anthraquinone-1,5-disulphonic acid disodium salt
分子式: C₁₄H₈O₈S₂
分子量: 412,29 g/mol
MDL编号: MFCD00044659
CAS号: 853-35-0
EINECS: 212-716-2

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR