D-天冬酰胺 ≥99.0% (通过HPLC,滴定分析)
供应商: TCI EUROPE
Synonyms:
(R)-2-氨基琥珀酸-4-酰胺
, (R)-(-)-2-氨基琥珀酰胺酸
, (D)-天冬氨酸-4-酰胺
TCIAA0543-25GEA
0
CNY
TCIAA0543-25G
D-天冬酰胺 ≥99.0% (通过HPLC,滴定分析)
D-天冬酰胺
分子式:
C₄H₁₀N₂O₄ 分子量: 150,14 g/mol (132,12 anhy) |
MDL编号:
MFCD00149558 CAS号: 5794-24-1 EINECS: 218-163-3 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Optical purity (LC) | min. 98.0 ee% |
Purity (Nonaqueous Titration) | min. 99.0 % (calcd.on dried substance) |
Drying loss | 11.5 to 12.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...