8-氮杂腺嘌呤 ≥98.0% (通过HPLC)

供应商: TCI EUROPE

Synonyms: 8-氮杂-6-氨基嘌呤6-氨基-8-氮杂嘌呤

TCIAA0552-1GEA 0 CNY
TCIAA0552-1G
8-氮杂腺嘌呤 ≥98.0% (通过HPLC)
8-氮杂腺嘌呤
分子式: C₄H₄N₆
分子量: 136,12 g/mol
溶点: 320°C
MDL编号: MFCD00005697
CAS号: 1123-54-2
EINECS: 214-375-5

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR