6-Azathymine ≥98.0% (通过HPLC,滴定分析)
供应商: TCI EUROPE
TCIAA0556-1GEA
430
CNY
TCIAA0556-1G
6-Azathymine ≥98.0% (通过HPLC,滴定分析)
6-Azathymine
分子式:
C₄H₅N₃O₂ |
MDL编号:
MFCD00006457 CAS号: 932-53-6 EINECS: 213-253-9 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 212.0 to 216.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...