Azo rubine

供应商: TCI EUROPE
Warning

TCIAA0580-25GEA 0 CNY
TCIAA0580-25G
Azo rubine
Azo rubine
分子式: C₂₀H₁₄N₂O₇S₂
分子量: 502,42 g/mol
MDL编号: MFCD00003978
CAS号: 3567-69-9
EINECS: 222-657-4

现在订购

产品技术规格

Appearance Dark red to Brown powder to crystal
Purity (HPLC) min. 95.0 area%
Lambda max. 514.0 to 518.0 nm (H2O)
Absorbance (E1%1cm) min. 230 (H2O, 514.0 to 518.0 nm)
Drying loss max. 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR