Acid red 27 ≥90.0% (通过UV/Vis)

供应商: TCI EUROPE

TCIAA0583-25GEA 0 CNY
TCIAA0583-25G
Acid red 27 ≥90.0% (通过UV/Vis)
Acid red 27
分子式: C₂₀H₁₄N₂O₁₀S₃
分子量: 604,46 g/mol
溶点: 220°C
MDL编号: MFCD00004076
CAS号: 915-67-3
EINECS: 213-022-2

现在订购

产品技术规格

Appearance Red to Dark red to Brown powder to crystal
Purity (UV-vis method) min. 90.0 % (after drying)
Drying loss max. 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR