1-金刚烷甲胺 ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
Synonyms:
1-氨甲基金刚烷
TCIAA0721-1GEA
1025
CNY
TCIAA0721-1G
TCIAA0721-5G
1-金刚烷甲胺 ≥98.0% (通过滴定分析)
1-金刚烷甲胺
分子式:
C₁₁H₁₉N 分子量: 165,28 g/mol 沸点: 97°C 闪点: 92°C |
MDL编号:
MFCD00074750 CAS号: 17768-41-1 EINECS: 241-752-1 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...