1-Acetamidoadamantane ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAA0737-25GEA 385 CNY
TCIAA0737-25G
1-Acetamidoadamantane ≥98.0% (通过GC)
1-Acetamidoadamantane
分子式: C₁₂H₁₉NO
分子量: 193,29 g/mol
MDL编号: MFCD00074730
CAS号: 880-52-4
EINECS: 212-914-9

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 151.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR