Barium acrylate monomer ≥95.0% (通过滴定分析)

供应商: TCI EUROPE
Danger

TCIAA0760-25GEA 0 CNY
TCIAA0760-25G
Barium acrylate monomer ≥95.0% (通过滴定分析)
Barium acrylate monomer
分子式: C₃H₄O₂
分子量: 279,44 g/mol
MDL编号: MFCD00067204
CAS号: 17989-90-1
UN号: 1564
ADR: 6.1,III

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Bromination Method) min. 95.0 %
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR