1-Aminononadecane ≥96.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAA0765-10GEA
0
CNY
TCIAA0765-10G
1-Aminononadecane ≥96.0% (通过GC)
1-Aminononadecane
分子式:
C₁₉H₄₁N 分子量: 283,54 g/mol 溶点: 53°C 存储条件: Refrigerator |
MDL编号:
MFCD00047944 CAS号: 14130-05-3 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...