2-Amino-4-chlorophenol-6-sulphonic acid ≥99.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE
Danger

TCIAA0773-25GEA 0 CNY
TCIAA0773-25G
2-Amino-4-chlorophenol-6-sulphonic acid ≥99.0% (通过HPLC,滴定分析)
2-Amino-4-chlorophenol-6-sulphonic acid
分子式: C₆H₆ClNO₄S
分子量: 223,63 g/mol
MDL编号: MFCD00035768
CAS号: 88-23-3
EINECS: 201-813-5
UN号: 2811
ADR: 6.1,III

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 99.0 %
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR