2,3-Dibromopropyl acrylate ≥85.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAA0910-25GEA 1100 CNY
TCIAA0910-25G
2,3-Dibromopropyl acrylate ≥85.0% (通过GC)
2,3-Dibromopropyl acrylate
分子式: C₆H₈Br₂O₂
分子量: 271,94 g/mol
沸点: 105°C
MDL编号: MFCD00059461
CAS号: 19660-16-3
EINECS: 243-208-9

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 85.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR